3-(Trifluoromethylthio)phenol 95%

Supplier: Ambeed
Warning

Synonyms: 3-[(Trifluoromethyl)thio]phenol

A350456-1G A350456-250MG A350456-5G
77154-986EA 75 USD
77154-986 77154-988 77901-850
3-(Trifluoromethylthio)phenol 95%
3-(Trifluoromethylthio)phenol

Precaución: for Research Use Only

Formula: C₇H₅F₃OS
MDL Number: MFCD06248888
CAS Number: 3823-40-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR