m-PEG4-NHS ester 97%

Supplier: Ambeed
Warning

Synonyms: mPEG4-NHS Ester, Methyl-PEG4-NHS Ester, N-Succinimidyl 4,7,10,13-Tetraoxatetradecanoate, 2,5-Dioxopyrrolidin-1-yl 2,5,8,11-tetraoxatetradecan-14-oate, 4,7,10,13-Tetraoxatetradecanoic acid N-succinimidyl ester

A349766-250MG A349766-5G
77154-978EA 82.4 USD
77154-978 77685-994
m-PEG4-NHS ester 97%
m-PEG4-NHS ester

Precaución: for Research Use Only

Formula: C₁₄H₂₃NO₈
MW: 333.34 g/mol
MDL Number: MFCD11041112
CAS Number: 622405-78-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR