4-Bromoisoquinoline-8-carboxylic acid 95%

Supplier: Ambeed
Warning

A349657-1G A349657-250MG
77154-974EA 762 USD
77154-974 77154-976
4-Bromoisoquinoline-8-carboxylic acid 95%
4-Bromoisoquinoline-8-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₆BrNO₂
MDL Number: MFCD28133465
CAS Number: 1823254-04-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR