4-(2-Methylpropoxy)benzonitrile 98%

Supplier: Ambeed
Warning

A349491-1G A349491-250MG A349491-5G
77154-954EA 28 USD
77154-954 77154-956 77612-996
4-(2-Methylpropoxy)benzonitrile 98%
4-(2-Methylpropoxy)benzonitrile

Precaución: for Research Use Only

Formula: C₁₁H₁₃NO
MDL Number: MFCD08274895
CAS Number: 5203-15-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR