Phenazine-1,6-diol 95%

Supplier: Ambeed
Warning

A349031-100MG A349031-250MG
77154-950EA 64 USD
77154-950 77154-952
Phenazine-1,6-diol 95%
Phenazine-1,6-diol

Precaución: for Research Use Only

Formula: C₁₂H₈N₂O₂
MDL Number: MFCD01708477
CAS Number: 69-48-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR