Ethyl (2-bromophenyl)acetate 98%

Supplier: Ambeed
Warning

A348389-10G A348389-1G A348389-25G A348389-5G
77154-942EA 41.2 USD
77154-942 77154-944 77154-946 77154-948
Ethyl (2-bromophenyl)acetate 98%
Ethyl (2-bromophenyl)acetate

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrO₂
MW: 243.10 g/mol
MDL Number: MFCD00051921
CAS Number: 2178-24-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR