Z-Pyr-OtBu 97%

Supplier: Ambeed
Warning

A346058-1G A346058-250MG A346058-25G A346058-5G
77154-934EA 10 USD
77154-934 77154-936 77154-938 77154-940
Z-Pyr-OtBu 97%
Z-Pyr-OtBu

Precaución: for Research Use Only

Formula: C₁₇H₂₁NO₅
MDL Number: MFCD11975830
CAS Number: 81470-51-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR