Ondansetron hydrochloride dihydrate 98%

Supplier: Ambeed
Danger

Synonyms: 9-Methyl-3-((2-methyl-1H-imidazol-1-yl)methyl)-1,2,3,9-tetrahydro-4H-carbazol-4-one hydrochloride dihydrate, GR 3832 HCl 2H2O, 1,2,3,9-Tetrahydro-9-methyl-3-[(2-methyl-1H-imidazol-1-yl)methyl]-4H-carbazol-4-one Hydrochloride, SN-37 HCl 2H2O, NSC665799 HCl 2H2O

A344767-100MG A344767-10MG A344767-1G A344767-25G A344767-50MG A344767-5G
77154-922EA 10.3 USD
77154-922 77154-924 77154-926 77154-928 77154-930 77154-932
Ondansetron hydrochloride dihydrate 98%
Ondansetron hydrochloride dihydrate

Precaución: for Research Use Only

Formula: C₁₈H₁₉N₃O·HCl·₂H₂O
MW: 365.86 g/mol
MDL Number: MFCD00374371
CAS Number: 103639-04-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR