Pentafluorophenylhydrazine 98%
Supplier: Ambeed
|
Danger
|
A343991-100G
A343991-25G
A343991-5G
A343991-1G
77154-894EA
482.04
USD
77154-894
77154-896
77594-012
77685-984
Pentafluorophenylhydrazine 98%
Pentafluorophenylhydrazine
Precaución: for Research Use Only
Formula:
C₆H₃F₅N₂ MW: 198.1 g/mol |
MDL Number:
MFCD00007574 CAS Number: 828-73-9 UN: 2811 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...