5-Chloropyrazolo[1,5-a]pyrimidin-3-amine 97%

Supplier: Ambeed
Warning

A341476-100MG A341476-1G A341476-250MG
77154-876EA 35.02 USD
77154-876 77154-878 77154-880
5-Chloropyrazolo[1,5-a]pyrimidin-3-amine 97%
5-Chloropyrazolo[1,5-a]pyrimidin-3-amine

Precaución: for Research Use Only

Formula: C₆H₅ClN₄
MW: 168.584 g/mol
MDL Number: MFCD17016123
CAS Number: 1234616-50-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR