3-Iodo-1H-pyrazolo[3,4-b]pyridine-5-carbonitrile 95%

Supplier: Ambeed
Warning

A337854-100MG A337854-1G
77154-858EA 48.41 USD
77154-858 77154-860
3-Iodo-1H-pyrazolo[3,4-b]pyridine-5-carbonitrile 95%
3-Iodo-1H-pyrazolo[3,4-b]pyridine-5-carbonitrile

Precaución: for Research Use Only

Formula: C₇H₃IN₄
MW: 270.03 g/mol
MDL Number: MFCD23098186
CAS Number: 1638764-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR