3-Chloro-4-(trifluoromethyl)benzoic acid 98%

Supplier: Ambeed
Warning

A337483-100G A337483-10G A337483-1G A337483-250MG A337483-25G A337483-5G
77154-832EA 203 USD
77154-832 77154-834 77154-836 77154-838 77154-840 77154-842
3-Chloro-4-(trifluoromethyl)benzoic acid 98%
3-Chloro-4-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₄ClF₃O₂
MDL Number: MFCD04972755
CAS Number: 115754-20-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR