3-Bromo-5-chloro-2-hydroxybenzyl alcohol 95%

Supplier: Ambeed
Warning

A337423-1G A337423-250MG
77154-828EA 74.16 USD
77154-828 77154-830
3-Bromo-5-chloro-2-hydroxybenzyl alcohol 95%
3-Bromo-5-chloro-2-hydroxybenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₆BrClO₂
MW: 237.48 g/mol
MDL Number: MFCD06202648
CAS Number: 116795-47-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR