Tasquinimod 98%

Supplier: Ambeed
Warning

Synonyms: 4-Hydroxy-5-methoxy-N,1-dimethyl-2-oxo-N-(4-(trifluoromethyl)phenyl)-1,2-dihydroquinoline-3-carboxamide

A337216-10MG A337216-1MG A337216-25MG A337216-2MG A337216-5MG
77154-818EA 152.44 USD
77154-818 77154-820 77154-822 77154-824 77154-826
Tasquinimod 98%
Tasquinimod

Precaución: for Research Use Only

Formula: C₂₀H₁₇F₃N₂O₄
MW: 406.36 g/mol
MDL Number: MFCD18251454
CAS Number: 254964-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR