Pyridine-2-carbaldehyde 98%

Supplier: Ambeed
Danger

Synonyms: 2-Pyridinecarboxaldehyde, 2-Pyridinecarbaldehyde, Pyridine-2-carboxaldehyde

A336992-25G A336992-5G A336992-10G A336992-100G
77154-814EA 14 USD
77154-814 77154-816 77550-968 77685-964
Pyridine-2-carbaldehyde 98%
Pyridine-2-carbaldehyde

Precaución: for Research Use Only

Formula: C₆H₅NO
MDL Number: MFCD00006290
CAS Number: 1121-60-4
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR