Z-Gly-Pro 97%

Supplier: Ambeed
Warning

Synonyms: (S)-1-(2-(((Benzyloxy)carbonyl)amino)acetyl)pyrrolidine-2-carboxylic acid, Z-Gly-Pro-OH

A336496-1G A336496-250MG A336496-5G A336496-10G
77154-802EA 22.66 USD
77154-802 77154-804 77154-806 77612-932
Z-Gly-Pro 97%
Z-Gly-Pro

Precaución: for Research Use Only

Formula: C₁₅H₁₈N₂O₅
MW: 306.318 g/mol
MDL Number: MFCD00037341
CAS Number: 1160-54-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR