(3R,4S)-4-Aminotetrahydro-2H-pyran-3-ol hydrochloride 98%

Supplier: Ambeed
Warning

A334492-100MG A334492-250MG
77154-780EA 108.15 USD
77154-780 77154-782
(3R,4S)-4-Aminotetrahydro-2H-pyran-3-ol hydrochloride 98%
(3R,4S)-4-Aminotetrahydro-2H-pyran-3-ol hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₂ClNO₂
MW: 153.60 g/mol
MDL Number: MFCD23159220
CAS Number: 1096594-11-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR