2',6'-Difluoroacetophenone 97%

Supplier: Ambeed
Warning

A334277-100G A334277-10G A334277-1G A334277-25G A334277-5G
77154-764EA 167 USD
77154-764 77154-766 77154-768 77154-770 77154-772
2',6'-Difluoroacetophenone 97%
2',6'-Difluoroacetophenone

Precaución: for Research Use Only

Formula: C₈H₆F₂O
MDL Number: MFCD00000328
CAS Number: 13670-99-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR