2-Fluoro-5-(methoxy(methyl)carbamoyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: N-Methoxy-N-methyl 3-borono-4-fluorobenzamide

A333998-1G A333998-250MG A333998-5G
77154-758EA 166 USD
77154-758 77154-760 77154-762
2-Fluoro-5-(methoxy(methyl)carbamoyl)phenylboronic acid 98%
2-Fluoro-5-(methoxy(methyl)carbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BFNO₄
MDL Number: MFCD08436038
CAS Number: 874289-59-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR