2-Oxa-5-azabicyclo[2.2.2]octane hemioxalate 95%

Supplier: Ambeed
Warning

A333911-100MG A333911-1G A333911-250MG
77154-752EA 113.3 USD
77154-752 77154-754 77154-756
2-Oxa-5-azabicyclo[2.2.2]octane hemioxalate 95%
2-Oxa-5-azabicyclo[2.2.2]octane hemioxalate

Precaución: for Research Use Only

Formula: C₁₄H₂₄N₂O₆
MW: 316.35 g/mol
MDL Number: MFCD28100842
CAS Number: 1523606-41-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR