Ethyl 4-methyloxazole-2-carboxylate 96%
Supplier: Ambeed
|
Warning
|
A333612-100MG
A333612-1G
A333612-250MG
A333612-5G
77154-744EA
96.82
USD
77154-744
77154-746
77154-748
77154-750
Ethyl 4-methyloxazole-2-carboxylate 96%
Ethyl 4-methyloxazole-2-carboxylate
Precaución: for Research Use Only
Formula:
C₇H₉NO₃ MW: 155.15 g/mol |
MDL Number:
MFCD09038239 CAS Number: 90892-99-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...