(6-Chloro-2-fluoro-3-methoxyphenyl)acetic acid 98+%

Supplier: Ambeed
Warning

A332686-1G A332686-250MG A332686-5G
77154-734EA 56 USD
77154-734 77901-752 77900-698
(6-Chloro-2-fluoro-3-methoxyphenyl)acetic acid 98+%
(6-Chloro-2-fluoro-3-methoxyphenyl)acetic acid

Precaución: for Research Use Only

Formula: C₉H₈ClFO₃
MDL Number: MFCD09832273
CAS Number: 1017777-83-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR