3'-Aminobiphenyl-3-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3'-Amino-[1,1'-biphenyl]-3-carboxylic acid

A332343-250MG A332343-5G
77154-730EA 38.11 USD
77154-730 77154-732
3'-Aminobiphenyl-3-carboxylic acid 97%
3'-Aminobiphenyl-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₁NO₂
MW: 213.24 g/mol
MDL Number: MFCD03839954
CAS Number: 124221-71-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR