ZM-306416 98%

Supplier: Ambeed
Warning

Synonyms: N-(4-Chloro-2-fluorophenyl)-6,7-dimethoxyquinazolin-4-amine, CB-676475

A331607-100MG A331607-10MG A331607-1MG A331607-50MG A331607-5MG A331607-250MG
77154-714EA 350.2 USD
77154-714 77154-716 77154-718 77154-720 77154-722 77612-904
ZM-306416 98%
ZM-306416

Precaución: for Research Use Only

Formula: C₁₆H₁₃ClFN₃O₂
MW: 333.747 g/mol
MDL Number: MFCD02683969
CAS Number: 690206-97-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR