2,3-Difluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol 98%

Supplier: Ambeed
Warning

A331597-1G A331597-250MG
77154-710EA 326.51 USD
77154-710 77154-712
2,3-Difluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol 98%
2,3-Difluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol

Precaución: for Research Use Only

Formula: C₁₂H₁₅BF₂O₃
MW: 256.05 g/mol
MDL Number: MFCD16996287
CAS Number: 1220219-43-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR