2-Fluoro-N-methylbenzamide 95%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-n-methylbenzamide

A331502-1G A331502-250MG A331502-5G A331502-25G
77154-704EA 35 USD
77154-704 77154-706 77154-708 77901-795
2-Fluoro-N-methylbenzamide 95%
2-Fluoro-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₈H₈FNO
MW: 153.16 g/mol
MDL Number: MFCD01214098
CAS Number: 52833-63-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR