3-(Chloromethyl)-5-fluoropyridine hydrochloride 97%

Supplier: Ambeed
Danger

A330897-100MG A330897-1G A330897-250MG A330897-25G A330897-5G
77154-694EA 32 USD
77154-694 77154-696 77154-698 77892-683 77892-684
3-(Chloromethyl)-5-fluoropyridine hydrochloride 97%
3-(Chloromethyl)-5-fluoropyridine hydrochloride

Precaución: for Research Use Only

Formula: C₆H₆Cl₂FN
MDL Number: MFCD19687170
CAS Number: 1222556-83-3
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR