(R)-(+)-ɑ-Amino-γ-butyrolactone hydrochloride 97%

Supplier: Ambeed
Warning

A329271-10G A329271-1G A329271-25G A329271-5G
77154-668EA 5.15 USD
77154-668 77154-670 77154-672 77154-674 77154-676 77154-678
(R)-(+)-ɑ-Amino-γ-butyrolactone hydrochloride 97%
(R)-(+)-alpha-Amino-gamma-butyrolactone hydrochloride

Precaución: for Research Use Only

Formula: C₄H₇NO₂·HCl
MDL Number: MFCD00674071
CAS Number: 104347-13-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR