(R)-tert-Butyl 3-(4-amino-3-iodo-1H-pyrazolo[3,4-d]pyrimidin-1-yl)piperidine-1-carboxylate 95%

Supplier: Ambeed
Warning

A328553-100MG A328553-1G A328553-250MG A328553-5G
77154-660EA 18.54 USD
77154-660 77154-662 77154-664 77154-666
(R)-tert-Butyl 3-(4-amino-3-iodo-1H-pyrazolo[3,4-d]pyrimidin-1-yl)piperidine-1-carboxylate 95%
(R)-tert-Butyl 3-(4-amino-3-iodo-1H-pyrazolo[3,4-d]pyrimidin-1-yl)piperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₅H₂₁IN₆O₂
MW: 444.27 g/mol
MDL Number: MFCD27987084
CAS Number: 1276110-38-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR