ANA-12 98%

Supplier: Ambeed
Warning

Synonyms: N-(2-((2-oxoazepan-3-yl)carbamoyl)phenyl)benzo[b]thiophene-2-carboxamide, N-[2-[[(Hexahydro-2-oxo-1H-azepin-3-yl)amino]carbonyl]phenyl]benzo[b]thiophene-2-carboxamide, N-[2-[(2-oxoazepan-3-yl)carbamoyl]phenyl]-1-benzothiophene-2-carboxamide

A327589-10MG A327589-1MG A327589-50MG A327589-5MG
77154-642EA 88.58 USD
77154-642 77154-644 77154-646 77154-648
ANA-12 98%
ANA-12

Precaución: for Research Use Only

Formula: C₂₂H₂₁N₃O₃S
MW: 407.49 g/mol
MDL Number: MFCD00117444
CAS Number: 219766-25-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR