4-(Phenoxymethyl)benzeneboronic acid pinacol ester 98+%

Supplier: Ambeed
Warning

Synonyms: 4,4,5,5-Tetramethyl-2-(4-(phenoxymethyl)phenyl)-1,3,2-dioxaborolane

A327172-1G A327172-250MG A327172-5G
77154-626EA 40 USD
77154-626 77154-628 77154-630
4-(Phenoxymethyl)benzeneboronic acid pinacol ester 98+%
4-(Phenoxymethyl)benzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₉H₂₃BO₃
MDL Number: MFCD18827485
CAS Number: 946409-21-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR