Potassium 3-formyl-4-methoxyphenyltrifluoroborate 95+%

Supplier: Ambeed
Warning

A324684-1G A324684-250MG A324684-5G
77154-584EA 28.84 USD
77154-584 77154-586 77154-588
Potassium 3-formyl-4-methoxyphenyltrifluoroborate 95+%
Potassium 3-formyl-4-methoxyphenyltrifluoroborate

Precaución: for Research Use Only

Formula: C₈H₇BF₃KO₂
MW: 242.04 g/mol
MDL Number: MFCD09993570
CAS Number: 1393683-77-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR