GSK-650394 98%

Supplier: Ambeed
Warning

Synonyms: 2-Cyclopentyl-4-(5-phenyl-1H-pyrrolo[2,3-b]pyridin-3-yl)benzoic acid

A324075-10MG A324075-1MG A324075-50MG A324075-5MG
77154-572EA 76 USD
77154-572 77154-574 77154-576 77154-578
GSK-650394 98%
GSK-650394

Precaución: for Research Use Only

Formula: C₂₅H₂₂N₂O₂
MDL Number: MFCD12828779
CAS Number: 890842-28-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR