Boc-(2S,4S)-4-amino-1-Fmoc-pyrrolidine-2-carboxylic Acid 95%

Supplier: Ambeed
Warning

A323311-100MG A323311-1G A323311-250MG A323311-5G
77154-556EA 40.17 USD
77154-556 77154-558 77154-560 77154-562
Boc-(2S,4S)-4-amino-1-Fmoc-pyrrolidine-2-carboxylic Acid 95%
Boc-(2S,4S)-4-amino-1-Fmoc-pyrrolidine-2-carboxylic Acid

Precaución: for Research Use Only

Formula: C₂₅H₂₈N₂O₆
MW: 452.50 g/mol
MDL Number: MFCD02179641
CAS Number: 221352-74-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR