UNC-2025 98+%
Supplier: Ambeed
|
Warning
|
Synonyms:
MRX-6313, (1R,4R)-4-(2-(butylamino)-5-(4-((4-methylpiperazin-1-yl)methyl)phenyl)-7H-pyrrolo[2,3-d]pyrimidin-7-yl)cyclohexanol, UNC2025, trans-4-(2-(Butylamino)-5-(4-((4-methylpiperazin-1-yl)methyl)phenyl)-7H-pyrrolo[2,3-d]pyrimidin-7-yl)cyclohexanol
A321872-10MG
A321872-1MG
A321872-2MG
A321872-50MG
A321872-5MG
A321872-100MG
77154-536EA
26
USD
77154-536
77154-538
77154-540
77154-542
77154-544
77685-921
UNC-2025 98+%
UNC-2025
Precaución: for Research Use Only
Formula:
C₂₈H₄₀N₆O MW: 476.66 g/mol |
MDL Number:
MFCD28142874 CAS Number: 1429881-91-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...