1,3-Difluoro-4-nitrobenzene 98%
Supplier: Ambeed
|
Danger
|
A321735-100G
A321735-25G
A321735-500G
A321735-10G
77154-518EA
12
USD
77154-518
77154-520
77154-522
77901-915
1,3-Difluoro-4-nitrobenzene 98%
1,3-Difluoro-4-nitrobenzene
Precaución: for Research Use Only
Formula:
C₆H₃F₂NO₂ |
MDL Number:
MFCD00007050 CAS Number: 446-35-5 UN: 2810 ADR: 6.1,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...