tert-Butyl 2-(Phenylmethylsulfonamido)acetate 95%

Supplier: Ambeed
Warning

A320561-100MG A320561-1G A320561-250MG
77154-504EA 28.84 USD
77154-504 77154-506 77154-508
tert-Butyl 2-(Phenylmethylsulfonamido)acetate 95%
tert-Butyl 2-(Phenylmethylsulfonamido)acetate

Precaución: for Research Use Only

Formula: C₁₃H₁₉NO₄S
MW: 285.36 g/mol
MDL Number: MFCD09998226
CAS Number: 1015856-04-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR