2-Bromo-5-nitrobenzonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 1-Cyano-2-bromo-5-nitrobenzene

A319052-10G A319052-1G A319052-5G
77154-478EA 50.47 USD
77154-478 77154-480 77154-482
2-Bromo-5-nitrobenzonitrile 97%
2-Bromo-5-nitrobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃BrN₂O₂
MW: 227.02 g/mol
MDL Number: MFCD00234249
CAS Number: 134604-07-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR