2,3-Dihydroxy-5-nitrobenzaldehyde 98%
Supplier: Ambeed
|
Warning
|
A317047-100MG
A317047-250MG
A317047-5G
A317047-1G
77154-464EA
66
USD
77154-464
77154-466
77154-468
77901-475
2,3-Dihydroxy-5-nitrobenzaldehyde 98%
2,3-Dihydroxy-5-nitrobenzaldehyde
Precaución: for Research Use Only
Formula:
C₇H₅NO₅ |
MDL Number:
MFCD25964229 CAS Number: 52924-54-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...