1,3-Dimethyl-2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-carboxylic acid 98%

Supplier: Ambeed
Warning

A316130-100MG A316130-1G A316130-250MG A316130-5G
77154-456EA 6 USD
77154-456 77154-458 77154-460 77154-462
1,3-Dimethyl-2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-carboxylic acid 98%
1,3-Dimethyl-2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₈N₂O₄
MDL Number: MFCD09901808
CAS Number: 4869-45-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR