trans-3-Methoxycyclobutanamine hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A315906-100MG
A315906-1G
77154-452EA
66.95
USD
77154-452
77154-454
trans-3-Methoxycyclobutanamine hydrochloride 97%
trans-3-Methoxycyclobutanamine hydrochloride
Precaución: for Research Use Only
Formula:
C₅H₁₂ClNO |
MDL Number:
MFCD22415180 CAS Number: 1408074-49-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...