H-Cys(Trt)-OtBu·HCl 95%

Supplier: Ambeed
Warning

Synonyms: H-Cys(Trt)-OtBu.HCl

A313636-1G A313636-250MG A313636-25G A313636-5G
77154-430EA 70.04 USD
77154-430 77154-432 77154-434 77154-436
H-Cys(Trt)-OtBu·HCl 95%
H-Cys(Trt)-OtBu·HCl

Precaución: for Research Use Only

Formula: C₂₆H₂₉NO₂S·HCl
MW: 456 g/mol
MDL Number: MFCD21363202
CAS Number: 158009-03-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR