6-(Tritylthio)hexanoic acid 95%
Supplier: Ambeed
|
Warning
|
A313603-1G
A313603-250MG
A313603-100MG
A313603-5G
77154-428EA
258
USD
77154-428
77612-808
77685-890
77685-891
6-(Tritylthio)hexanoic acid 95%
6-(Tritylthio)hexanoic acid
Precaución: for Research Use Only
Formula:
C₂₅H₂₆O₂S |
MDL Number:
MFCD11226801 CAS Number: 80441-55-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...