1-Methyl-3-octylimidazolium tetrafluoroborate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-Methyl-3-n-octylimidazolium Tetrafluoroborate, 1-octyl-3-methylimidazolium tetrafluoroborate, [OMIM] [BF4]
A313422-100G
A313422-25G
77154-424EA
117.42
USD
77154-424
77154-426
1-Methyl-3-octylimidazolium tetrafluoroborate 97%
1-Methyl-3-octylimidazolium tetrafluoroborate
Precaución: for Research Use Only
Formula:
C₁₂H₂₃BF₄N₂ MW: 282.13 g/mol |
MDL Number:
MFCD03095434 CAS Number: 244193-52-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...