AC-Tyr(tBu)-OH 97%

Supplier: Ambeed
Warning

A313070-1G A313070-250MG
77154-416EA 20 USD
77154-416 77154-418
AC-Tyr(tBu)-OH 97%
AC-Tyr(tBu)-OH

Precaución: for Research Use Only

Formula: C₁₅H₂₁NO₄
MDL Number: MFCD00236772
CAS Number: 201292-99-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR