D-Phe-OEt HCl 98%

Supplier: Ambeed
Warning

Synonyms: (R)-Ethyl 2-amino-3-phenylpropanoate hydrochloride

A313054-100G A313054-25G A313054-5G
77154-410EA 109.18 USD
77154-410 77154-412 77154-414
D-Phe-OEt HCl 98%
D-Phe-OEt HCl

Precaución: for Research Use Only

Formula: C₁₁H₁₅NO₂·HCl
MW: 229.70 g/mol
MDL Number: MFCD00198196
CAS Number: 63060-94-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR