1-(3-Fluoropyridin-4-yl)ethanol 95%

Supplier: Ambeed
Warning

A312744-1G A312744-250MG
77154-388EA 31.93 USD
77154-388 77154-390 77154-392
1-(3-Fluoropyridin-4-yl)ethanol 95%
1-(3-Fluoropyridin-4-yl)ethanol

Precaución: for Research Use Only

Formula: C₇H₈FNO
MDL Number: MFCD18384616
CAS Number: 87674-15-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR