Diphenylammonium trifluoromethanesulfonate 98%

Supplier: Ambeed
Warning

Synonyms: Diphenylammonium Trifluoromethanesulphonate, DPAT

A312291-1G A312291-5G A312291-25G
77154-386EA 17 USD
77154-386 77901-901 77901-239
Diphenylammonium trifluoromethanesulfonate 98%
Diphenylammonium trifluoromethanesulfonate

Precaución: for Research Use Only

Formula: C₁₃H₁₂F₃NO₃S
MW: 319.30 g/mol
MDL Number: MFCD11042671
CAS Number: 164411-06-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR