2-Bromo-6-(trifluoromethyl)pyridine 98%

Supplier: Ambeed
Danger

A311139-10G A311139-1G A311139-25G A311139-5G
77154-376EA 11 USD
77154-376 77154-378 77154-380 77154-382
2-Bromo-6-(trifluoromethyl)pyridine 98%
2-Bromo-6-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₃BrF₃N
MW: 226
MDL Number: MFCD00153087
CAS Number: 189278-27-1
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR