3-Chloro-2,4,5-trifluorobenzoyl chloride 95%

Supplier: Ambeed
Danger

A310914-10G A310914-1G A310914-25G
77154-370EA 73.13 USD
77154-370 77154-372 77154-374
3-Chloro-2,4,5-trifluorobenzoyl chloride 95%
3-Chloro-2,4,5-trifluorobenzoyl chloride

Precaución: for Research Use Only

Formula: C₇HCl₂F₃O
MW: 228.983 g/mol
MDL Number: MFCD04039245
CAS Number: 101513-78-4
UN: 1760
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR